siRNA Details
Gene Symbol: 4930429D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015233.1 | - | 1, 2 | 71 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714787.1 | - | 1, 2 | 68 | Chr. 5 - 103231340 - 103299027 [-] on Build GRCm38 |
