siRNA Details
| Gene Symbol: 4930429C20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015232.1 | - | 1 | 225 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714786.1 | - | 1 | 225 | Chr. 14 - 71421012 - 71421613 [+] on Build GRCm38 | |

4930429C20Rik
Genomic Map