siRNA Details
| Gene Symbol: 4930429F11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033463.1 | 1 | 99 | |||
| GenBank mRNA | AK015235.1 | - | 1 | 100 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714790.1 | - | 1 | 100 | Chr. 17 - 79476593 - 79477885 [-] on Build GRCm38 | |

4930429F11Rik
Genomic Map