siRNA Details
| Gene Symbol: 4930429F11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033463.1 | 2 | 1071 | |||
| GenBank mRNA | AK015235.1 | - | 2 | 1072 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930429F11Rik| Gene Symbol: 4930429F11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033463.1 | 2 | 1071 | |||
| GenBank mRNA | AK015235.1 | - | 2 | 1072 | ||