siRNA Details
Gene Symbol: 4930429E23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015234.1 | - | 581 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714789.1 | - | 1 | 577 | Chr. 4 - 133910794 - 133911443 [+] on Build GRCm38 |
