siRNA Details
| Gene Symbol: 4930429E23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015234.1 | - | 484 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714789.1 | - | 1 | 480 | Chr. 4 - 133910794 - 133911443 [+] on Build GRCm38 | |

4930429E23Rik
Genomic Map