siRNA Details
Gene Symbol: 4930429F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040734.1 | 3 | 981 | |||
GenBank mRNA | AK015237.1 | - | 3 | 981 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930429F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040734.1 | 3 | 981 | |||
GenBank mRNA | AK015237.1 | - | 3 | 981 |