siRNA Details
| Gene Symbol: 4930429F24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040734.1 | 3 | 1338 | |||
| GenBank mRNA | AK015237.1 | - | 3 | 1338 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930429F24Rik| Gene Symbol: 4930429F24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040734.1 | 3 | 1338 | |||
| GenBank mRNA | AK015237.1 | - | 3 | 1338 | ||