siRNA Details
| Gene Symbol: 4930429F24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040734.1 | 3 | 387 | |||
| GenBank mRNA | AK015237.1 | - | 3 | 387 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714792.1 | - | 3 | 384 | Chr. 9 - 79793644 - 79802374 [+] on Build GRCm38 | |

4930429F24Rik
Genomic Map