siRNA Details
| Gene Symbol: 4930507D10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110449.1 | 3 | 774 | |||
| GenBank mRNA | AK015727.1 | - | 3 | 776 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930507D10Rik| Gene Symbol: 4930507D10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110449.1 | 3 | 774 | |||
| GenBank mRNA | AK015727.1 | - | 3 | 776 | ||