siRNA Details
| Gene Symbol: Mro | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015725.1 | - | 4 | 638 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715266.1 | - | 4 | 638 | Chr. 18 - 73859394 - 73881332 [+] on Build GRCm38 | |

Mro
Genomic Map