siRNA Details
| Gene Symbol: 4930507A01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015723.1 | - | 1 | 1337 | ||
| AK016354.1 | - | 1 | 1336 | |||
| AV265880.1 | - | 1 | 129 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930507A01Rik| Gene Symbol: 4930507A01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015723.1 | - | 1 | 1337 | ||
| AK016354.1 | - | 1 | 1336 | |||
| AV265880.1 | - | 1 | 129 | |||