siRNA Details
| Gene Symbol: Dzip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009865.1 | - | 1 | 477 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI729606.1 | - | 1 | 476 | Chr. 16 - 48993495 - 48994098 [-] on Build GRCm38 | |

Dzip3
Genomic Map