siRNA Details
| Gene Symbol: Fbxw27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007274.1 | - | 5 | 593 | ||
| BB702760.1 | - | 4 | 461 | |||
| CO804891.1 | - | 4 | 521 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxw27| Gene Symbol: Fbxw27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007274.1 | - | 5 | 593 | ||
| BB702760.1 | - | 4 | 461 | |||
| CO804891.1 | - | 4 | 521 | |||