siRNA Details
| Gene Symbol: 1700125G02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040651.1 | 2 | 391 | |||
| GenBank mRNA | AK007278.1 | - | 2 | 397 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707379.1 | - | 2 | 395 | Chr. 4 - 124890888 - 124894782 [-] on Build GRCm38 | |

1700125G02Rik
Genomic Map