siRNA Details
| Gene Symbol: 1700125G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040548.1 | 2 | 402 | |||
| GenBank mRNA | AK007279.1 | - | 2 | 402 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707381.1 | - | 409 | Chr. - - [-] on Build | ||

1700125G22Rik
Genomic Map