siRNA Details
| Gene Symbol: D230015J17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085005.1 | - | 1 | 772 | ||
| AK148836.1 | - | 1 | 1034 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D230015J17Rik| Gene Symbol: D230015J17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085005.1 | - | 1 | 772 | ||
| AK148836.1 | - | 1 | 1034 | |||