siRNA Details
| Gene Symbol: Gm26787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007186.1 | - | 4 | 441 | ||
| AK018988.1 | - | 4 | 420 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707377.1 | - | 4 | 420 | Chr. 9 - 36767910 - 36782677 [+] on Build GRCm38 | |

Gm26787
Genomic Map