siRNA Details
| Gene Symbol: Gm16861 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037666.1 | 3 | 1313 | |||
| GenBank mRNA | AK078615.2 | - | 3 | 1311 | ||
| BC061511.1 | - | 3 | 1361 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16861| Gene Symbol: Gm16861 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037666.1 | 3 | 1313 | |||
| GenBank mRNA | AK078615.2 | - | 3 | 1311 | ||
| BC061511.1 | - | 3 | 1361 | |||