siRNA Details
Gene Symbol: Cdc73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018410.1 | - | 1633 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX518593.1 | - | 1 | 133 | Chr. 1 - 143599034 - 143599540 [-] on Build GRCm38 |
