siRNA Details
| Gene Symbol: A930036K24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020935.1 | - | 1 | 1163 | ||
| BG297411.1 | - | 247 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK044545.1 | - | 1188 | Chr. - - [-] on Build | ||

A930036K24Rik
Genomic Map