siRNA Details
| Gene Symbol: 8430408J09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018398.1 | - | 1 | 702 | ||
| AK018417.1 | - | 1 | 702 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

8430408J09Rik| Gene Symbol: 8430408J09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018398.1 | - | 1 | 702 | ||
| AK018417.1 | - | 1 | 702 | |||