siRNA Details
| Gene Symbol: Celrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038008.1 | 5 | 1081 | |||
| NR_110450.1 | 3 | 820 | ||||
| GenBank mRNA | AK138346.1 | - | 5 | 1082 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Celrr| Gene Symbol: Celrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038008.1 | 5 | 1081 | |||
| NR_110450.1 | 3 | 820 | ||||
| GenBank mRNA | AK138346.1 | - | 5 | 1082 | ||