siRNA Details
Gene Symbol: 1700031P21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045910.1 | 3 | 273 | |||
GenBank mRNA | AK006587.1 | - | 3 | 274 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706590.1 | - | 3 | 269 | Chr. 12 - 52599984 - 52602318 [-] on Build GRCm38 |
