siRNA Details
| Gene Symbol: 1700031P21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045910.1 | 1 | 112 | |||
| GenBank mRNA | AK006587.1 | - | 1 | 113 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700031P21Rik| Gene Symbol: 1700031P21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045910.1 | 1 | 112 | |||
| GenBank mRNA | AK006587.1 | - | 1 | 113 | ||