siRNA Details
| Gene Symbol: 1700031P21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045910.1 | 1 | 134 | |||
| GenBank mRNA | AK006587.1 | - | 1 | 135 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 1700031P21Rik
1700031P21Rik| Gene Symbol: 1700031P21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045910.1 | 1 | 134 | |||
| GenBank mRNA | AK006587.1 | - | 1 | 135 | ||
 Back To Top
 Back To Top