siRNA Details
| Gene Symbol: 5730405N03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017498.1 | - | 188 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY717286.1 | - | 1 | 185 | Chr. 14 - 117903137 - 117904042 [+] on Build GRCm38 | |

5730405N03Rik
Genomic Map