siRNA Details
| Gene Symbol: A930001A20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040549.1 | 5 | 972 | |||
| NR_040550.1 | 4 | 757 | ||||
| GenBank mRNA | AK020793.1 | - | 5 | 952 | ||
| AK044583.1 | - | 4 | 758 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930001A20Rik| Gene Symbol: A930001A20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040549.1 | 5 | 972 | |||
| NR_040550.1 | 4 | 757 | ||||
| GenBank mRNA | AK020793.1 | - | 5 | 952 | ||
| AK044583.1 | - | 4 | 758 | |||