siRNA Details
| Gene Symbol: A930009E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020797.1 | - | 1 | 441 | ||
| AK020836.1 | - | 1 | 472 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930009E08Rik| Gene Symbol: A930009E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020797.1 | - | 1 | 441 | ||
| AK020836.1 | - | 1 | 472 | |||