siRNA Details
| Gene Symbol: A930009E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020797.1 | - | 1 | 596 | ||
| AK020836.1 | - | 1 | 627 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM936805.1 | - | 1 | 459 | Chr. 1 - 97637530 - 97638207 [-] on Build GRCm38 | |

A930009E08Rik
Genomic Map