siRNA Details
| Gene Symbol: A930009E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020797.1 | - | 1 | 125 | ||
| AK020836.1 | - | 1 | 156 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY742154.1 | - | 1 | 155 | Chr. 1 - 97637735 - 97638374 [-] on Build GRCm38 | |

A930009E08Rik
Genomic Map