siRNA Details
| Gene Symbol: A930002H02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020799.1 | - | 1 | 1033 | ||
| BB277126.1 | - | 1 | 77 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930002H02Rik| Gene Symbol: A930002H02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020799.1 | - | 1 | 1033 | ||
| BB277126.1 | - | 1 | 77 | |||