siRNA Details
| Gene Symbol: 2310043L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037994.1 | 3 | 561 | |||
| GenBank mRNA | AK009784.1 | - | 3 | 565 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310043L19Rik| Gene Symbol: 2310043L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037994.1 | 3 | 561 | |||
| GenBank mRNA | AK009784.1 | - | 3 | 565 | ||