siRNA Details
| Gene Symbol: 2310043L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037994.1 | 1, 2 | 237 | |||
| GenBank mRNA | AK009784.1 | - | 1, 2 | 241 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310043L19Rik| Gene Symbol: 2310043L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037994.1 | 1, 2 | 237 | |||
| GenBank mRNA | AK009784.1 | - | 1, 2 | 241 | ||