siRNA Details
| Gene Symbol: 1500012K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045812.1 | 2 | 255 | |||
| NR_045814.1 | 2 | 249 | ||||
| GenBank mRNA | AK005235.1 | - | 2 | 256 | ||
| AK035406.1 | - | 2 | 249 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1500012K07Rik| Gene Symbol: 1500012K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045812.1 | 2 | 255 | |||
| NR_045814.1 | 2 | 249 | ||||
| GenBank mRNA | AK005235.1 | - | 2 | 256 | ||
| AK035406.1 | - | 2 | 249 | |||