siRNA Details
| Gene Symbol: 1500012K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045812.1 | 2 | 282 | |||
| NR_045814.1 | 2 | 276 | ||||
| GenBank mRNA | AK005235.1 | - | 2 | 283 | ||
| AK035406.1 | - | 2 | 276 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY705414.1 | - | 2 | 280 | Chr. 7 - 75308698 - 75318485 [+] on Build GRCm38 | |

1500012K07Rik
Genomic Map