siRNA Details
| Gene Symbol: 2310034G01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040418.1 | 1 | 277 | |||
| GenBank mRNA | AK009611.1 | - | 1 | 244 | ||
| AK051784.1 | - | 1 | 264 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310034G01Rik| Gene Symbol: 2310034G01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040418.1 | 1 | 277 | |||
| GenBank mRNA | AK009611.1 | - | 1 | 244 | ||
| AK051784.1 | - | 1 | 264 | |||