siRNA Details
| Gene Symbol: 4930500F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045758.1 | 1 | 115 | |||
| GenBank mRNA | AK015658.1 | - | 1 | 115 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930500F04Rik| Gene Symbol: 4930500F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045758.1 | 1 | 115 | |||
| GenBank mRNA | AK015658.1 | - | 1 | 115 | ||