siRNA Details
| Gene Symbol: 4921524L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK161257.1 | - | 2895 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV257521.2 | - | 1 | 644 | Chr. 18 - 6640370 - 6641041 [+] on Build GRCm38 | |

4921524L21Rik
Genomic Map