siRNA Details
| Gene Symbol: Rnf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139182.1 | - | 354 | |||
| AK151082.1 | - | 1534 | ||||
| AK153482.1 | - | 1535 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf11| Gene Symbol: Rnf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139182.1 | - | 354 | |||
| AK151082.1 | - | 1534 | ||||
| AK153482.1 | - | 1535 | ||||