siRNA Details
| Gene Symbol: Gm4791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086549.1 | - | 3 | 516 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB663240.1 | - | 3 | 511 | Chr. 9 - 46998745 - 47003475 [+] on Build GRCm38 | |

Gm4791
Genomic Map