siRNA Details
| Gene Symbol: Gm17751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038012.1 | 5 | 946 | |||
| NR_038013.1 | 5 | 762 | ||||
| GenBank mRNA | AK054265.1 | - | 5 | 762 | ||
| AK162144.1 | - | 5 | 946 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm17751| Gene Symbol: Gm17751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038012.1 | 5 | 946 | |||
| NR_038013.1 | 5 | 762 | ||||
| GenBank mRNA | AK054265.1 | - | 5 | 762 | ||
| AK162144.1 | - | 5 | 946 | |||