siRNA Details
| Gene Symbol: 1110064A23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004365.1 | - | 1 | 229 | ||
| BC040515.1 | - | 1 | 1292 | |||
| BF180451.1 | - | 1 | 322 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110064A23Rik| Gene Symbol: 1110064A23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004365.1 | - | 1 | 229 | ||
| BC040515.1 | - | 1 | 1292 | |||
| BF180451.1 | - | 1 | 322 | |||