siRNA Details
| Gene Symbol: 4930470O06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015539.1 | - | 1 | 170 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715087.1 | - | 1 | 170 | Chr. 8 - 59825649 - 59844721 [+] on Build GRCm38 | |

4930470O06Rik
Genomic Map