siRNA Details
| Gene Symbol: 9330175M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045151.2 | 5 | 1143 | |||
| GenBank mRNA | AK034309.1 | - | 5 | 1143 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9330175M20Rik| Gene Symbol: 9330175M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045151.2 | 5 | 1143 | |||
| GenBank mRNA | AK034309.1 | - | 5 | 1143 | ||