siRNA Details
| Gene Symbol: 9330175M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045151.2 | 4 | 321 | |||
| GenBank mRNA | AK034309.1 | - | 4 | 322 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB626329.1 | - | 4 | 322 | Chr. 1 - 51220284 - 51333779 [-] on Build GRCm38 | |

9330175M20Rik
Genomic Map