siRNA Details
Gene Symbol: 9330175M20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045151.2 | 5 | 1240 | |||
GenBank mRNA | AK034309.1 | - | 5 | 1240 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9330175M20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045151.2 | 5 | 1240 | |||
GenBank mRNA | AK034309.1 | - | 5 | 1240 |