siRNA Details
| Gene Symbol: Kalrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK081504.1 | - | 2 | 2031 | ||
| AK086150.1 | - | 1 | 287 | |||
| AK089128.1 | - | 1 | 289 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kalrn| Gene Symbol: Kalrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK081504.1 | - | 2 | 2031 | ||
| AK086150.1 | - | 1 | 287 | |||
| AK089128.1 | - | 1 | 289 | |||