siRNA Details
| Gene Symbol: Kalrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086150.1 | - | 1 | 712 | ||
| AK089128.1 | - | 1 | 714 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CX763511.1 | - | 1 | 96 | Chr. 16 - 33974208 - 33974914 [-] on Build GRCm38 | |

Kalrn
Genomic Map