siRNA Details
| Gene Symbol: Gm15395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC048597.1 | - | 2 | 287 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU945707.1 | - | 2 | 290 | Chr. 8 - 117036494 - 117043621 [+] on Build GRCm38 | |

Gm15395
Genomic Map